Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALAS1 Recombinant Rabbit Monoclonal Antibody (SA70-04)
GREENER_CHOICE

Catalog No. PIMA531972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531972 Supplier Invitrogen™ Supplier No. MA531972
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5' phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALAS1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SA70-04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene ALAS1
Gene Accession No. P13196
Gene Alias 5'-aminolevulinate synthase 1; 5-aminolevulinate synthase 1; 5-aminolevulinate synthase, nonspecific, mitochondrial; 5-aminolevulinic acid synthase 1; ALAS; ALA-S; ALAS1; Alas-1; ALAS3; ALASH; Alas-h; ALAS-N; aminolevulinate synthase H; aminolevulinate, delta-, synthase 1; aminolevulinic acid synthase 1; delta-ALA synthase 1; delta-ALA synthetase; delta-aminolevulinate synthase 1; MIG4; migration-inducing protein 4; OK/SW-cl.121; succinyl-CoA: glycine C-succinyl transferase
Gene Symbols ALAS1
Host Species Rabbit
Immunogen Recombinant protein within Human Alas1 aa 510-640.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.