Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143885
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143885 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143885 Supplier Invitrogen™ Supplier No. PA5143885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, rat liver tissue, rat heart tissue, mouse liver tissue, mouse heart tissue. ICC/IF: PC-3 cell. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

ALAS2 specifies an erythroid-specific mitochondrially located enzyme. It catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia.The product of this gene specifies an erythroid-specific mitochondrially located enzyme. The encoded protein catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALAS2
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ALAS2
Gene Accession No. P08680, P22557, Q63147
Gene Alias 5'-aminolevulinate synthase 2; 5-aminolevulinate synthase 2; 5-aminolevulinate synthase, erythroid-specific, mitochondrial; 5-aminolevulinic acid synthase 2; ALAS; Alas2; Alas-2; alas2 protein; Alase; ALAS-E; Aminolevulinate synthase 2, delta; aminolevulinate synthase erythroid specific isoform; aminolevulinate, delta-, synthase 2; aminolevulinate, delta-, synthase 2 (sideroblastic/hypochromic anemia); aminolevulinate, delta-, synthetase 2; aminolevulinic acid synthase 2, erythroid; ANH1; ASB; cb1063; Delta-ALA synthase 2; delta-ALA synthetase; delta-ALA synthetase 2; delta-aminolevulinate synthase 2; erythroid aminolevulinate synthase; erythroid-specific ALAS; erythroid-specific delta-aminolevulinate synthase ALAS-E; heme biosynthetic enzyme; PRO2399; RP5-884M20.1; sau; sauternes; si:dkey-24f17.4; SIDBA1; XLDPP; XLEPP; XLSA
Gene Symbols ALAS2
Host Species Rabbit
Immunogen E.coli-derived human ALAS2/ASB recombinant protein (Position: M1-D190).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11656, 212, 25748
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.