Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121007 Supplier Invitrogen™ Supplier No. PA5121007
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Mouse liver, Mouse spleen, Mouse lung. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: VKTHQFLFGI GRCPILATQG PNCSQIHLKA TKAGGDSPSW AKGHCPFMLS ELQDGKSKIV QKAAPEVQED VKAFKTGNYV FSYDQFFRDK IMEKKQDHTY RVFKTVNRWA DAYPFAQHFS EASVASKDVS VWCSNDYLGM SRHPQVLQAT QETLQRHGAG AGGTRNISGT SKFHVELEQE LAELHQKDSA LLFSSCFVAN DSTLFTLAKI LPGCEIYSDA GNHASMIQGI RNSGAAKFVF RHNDPDHLKK LLEKSNPKIP KIVAFETVHS M.

ALAS2 specifies an erythroid-specific mitochondrially located enzyme. It catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia.The product of this gene specifies an erythroid-specific mitochondrially located enzyme. The encoded protein catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALAS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALAS2
Gene Accession No. P08680, P22557, Q63147
Gene Alias 5'-aminolevulinate synthase 2; 5-aminolevulinate synthase 2; 5-aminolevulinate synthase, erythroid-specific, mitochondrial; 5-aminolevulinic acid synthase 2; ALAS; Alas2; Alas-2; alas2 protein; Alase; ALAS-E; Aminolevulinate synthase 2, delta; aminolevulinate synthase erythroid specific isoform; aminolevulinate, delta-, synthase 2; aminolevulinate, delta-, synthase 2 (sideroblastic/hypochromic anemia); aminolevulinate, delta-, synthetase 2; aminolevulinic acid synthase 2, erythroid; ANH1; ASB; cb1063; Delta-ALA synthase 2; delta-ALA synthetase; delta-ALA synthetase 2; delta-aminolevulinate synthase 2; erythroid aminolevulinate synthase; erythroid-specific ALAS; erythroid-specific delta-aminolevulinate synthase ALAS-E; heme biosynthetic enzyme; PRO2399; RP5-884M20.1; sau; sauternes; si:dkey-24f17.4; SIDBA1; XLDPP; XLEPP; XLSA
Gene Symbols ALAS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-320 of human ALAS2 (NP_001033057.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11656, 212, 25748
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.