Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALB Mouse anti-Human, Clone: 12D12, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14098
Description
Mouse monoclonal antibody raised against human ALB.
- High specificity
- High reactivity
Specifications
ALB | |
Monoclonal | |
Unconjugated | |
ELISA Immunoprecipitation Radioimmunoassay The optimal working dilution should be determined by the end user. | |
P02768 | |
ALB | |
Human serum ALB. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
12D12 | |
Mouse monoclonal antibody raised against human ALB. | |
In 0.01 M potassium phosphate buffer, 2.5 mM KCl, 0.135 M NaCl (1 mg/mL BSA, 0.05% sodium azide). | |
DKFZp779N1935/PRO0883/PRO0903/PRO1341 | |
Mouse | |
Ion exchange chromatography purification | |
RUO | |
213 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction