Learn More
ALB Mouse anti-Human, Clone: OTI3B10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF507286

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough `produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.Specifications
ALB | |
Monoclonal | |
Unconjugated | |
ALB | |
HSA, PRO0883, PRO0903, PRO1341 | |
Mouse | |
Affinity Chromatography | |
RUO | |
213 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02768 | |
ALB | |
Human recombit protein fragment corresponding to amino acids 19-609 of human ALB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.