Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albendazole, 98+%

Catalog No. aah2592509
Click to view available options
Quantity:
10 g
50 g
100 g

CAS: 54965-21-8 | C12H15N3O2S | 265.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00083232
InChI Key HXHWSAZORRCQMX-UHFFFAOYSA-N
Synonym albendazole, albenza, eskazole, valbazen, zentel, albendazol, proftril, albendazolum, bilutac, zental
PubChem CID 2082
ChEBI CHEBI:16664
IUPAC Name methyl N-(6-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 208°C to 210°C
Quantity 10 g
Merck Index 14,210
Formula Weight 265.34
Percent Purity ≥98%
Chemical Name or Material Albendazole
Hazard Category H315-H319-H335-H360D
Hazard Statement GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 259-414-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.