Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Albendazole Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175040
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175040 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175040 Supplier Invitrogen™ Supplier No. PA175040
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 10 μg/mL. Sensitivity: 10 ng/mL Albendazole produces 70% inhibition in a competitive ELISA employing Albendazole polyclonal antibody.

Anthelmintics or anti-helminthics are a class of drugs that are effective against a range of intestinal parasitic worms (helminths). Parasitic helminths must maintain an appropriate feeding site. Nematodes and trematodes must actively ingest and move food through their digestive tracts to maintain an appropriate energy state; these together with reproductive processes require a well defined and developed neuromuscular coordination. Anthelmintic treatment is a multi-targeting system designed to interfere with the integrity of parasite cells. The pharmacologic basis of the treatment for helminths involves the targeting of neuromuscular coordination, or protective mechanisms against host immunity, which lead to starvation, paralysis, and expulsion of the parasite. The benzimidazole class of drugs were introduced in 1961 and interfere with the parasite's ATP pathway on a cellular level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Albendazole
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Sheep
Immunogen Albendazole(5-propylthio)-BTG.
Purification Method purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.