Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albendazole 98.0+%, TCI America™
SDP

Catalog No. A194325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A194325G 25 g
A19435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A194325G Supplier TCI America Supplier No. A194325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00083232
InChI Key HXHWSAZORRCQMX-UHFFFAOYSA-N
Synonym albendazole, albenza, eskazole, valbazen, zentel, albendazol, proftril, albendazolum, bilutac, zental
PubChem CID 2082
ChEBI CHEBI:16664
IUPAC Name methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 25 g
Formula Weight 265.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Albendazole
RTECSNumber FD1100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.