Learn More
Eisco™ Albite Mineral Specimen
SCI_ED
Catalog No. S26687 Shop All Eisco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S26687 10/Pk.
1 options
Catalog No. S26687 Supplier Eisco™ Supplier No. ESNG0064PK10
Add to Cart
Add to Cart
Albite Mineral Specimen ideal for use in lessons about the Mohs hardness scale.
  • Used as a hardness standard of 6 (Mohs scale)
  • Exhibits two planes of cleavage
  • Correlates well to Next Generation Science Standards (NGSS) Evidence Statements
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Albite