Learn More
Eisco™ Albite Specimen
SCI_ED
Catalog No. S39787 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S39787 1 Ea.
1 options
Catalog No. S39787 Supplier Eisco™ Supplier No. ESNG0064
Add to Cart
Add to Cart
Albite is a variety of plagioclase feldspar that is rich in sodium. With a glossy sheen, 2 planes of cleavage, and a standard hardness of 6, this mineral is a classic specimen for lessons in mineralogy and petrology. Useful when trying to distinguish from other feldspars.
  • Albite sample approximately 1 in. (3cm) in size
  • Used as a hardness standard of 6, and exhibits two planes of cleavage
  • ideal size for use in classroom identification labs and exercises
  • Great sample for use in lessons about the Mohs hardness scale
  • Specimen comes in clear labeled and barcoded bag for ideal storage
  • Correlates well to Next Generation Science Standards (NGSS) Evidence Statements
Warranty and Services
1 year limited manufacturer warranty.
Specifications
Product Type Mineral
Includes Albite
Quantity 1 Ea.
Specimen Size 3 cm
Weight (Metric) 17 g