Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Antibody (HSA20), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179049H
Description
Albumin Monoclonal specifically detects Albumin in Human, Bovine (Negative) samples. It is validated for Western Blot, ELISA.Specifications
Albumin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ALB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human serum albumin. | |
0.2 mL | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Human, Bovine (Negative) | |
Purified |
Western Blot, ELISA | |
HSA20 | |
Western Blot, ELISA | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity with other tested human proteins. Does not react with bovine serum albumin, egg white albumin or human alpha fetoprotein. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction