Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Antibody (HSA20), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179049
Description
Albumin Monoclonal specifically detects Albumin in Human, Bovine (Negative) samples. It is validated for Western Blot, ELISA.Specifications
Albumin | |
Monoclonal | |
Unconjugated | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity with other tested human proteins. Does not react with bovine serum albumin, egg white albumin or human alpha fetoprotein. | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, ELISA | |
HSA20 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
ALB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human serum albumin. | |
0.2 mg | |
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Human, Bovine (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction