Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB6005700.05ML
Description
Albumin Polyclonal specifically detects Albumin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Albumin | |
Polyclonal | |
Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
P02768 | |
ALB | |
Purified human albumin. | |
0.05 mL | |
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
PBS with BSA with 0.09% Sodium Azide | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
This reacts with human albumin. This is essentially free of other rabbit serum proteins. Specificity is determined by Ouchterlony Double Diffusion (ODD) and immunoelectrophoresis (IEP) versus human serum and human albumin. | |
Store at 4C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction