Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Albumin, for biochemistry, from chicken egg, powder

Catalog No. p-4258661
Click to view available options
Quantity:
50 g

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
• It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications
• BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
• It is used to stabilize enzymes in restriction digestions in molecular biology experiments
• It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
• BSA is a nutritive agent and used as a supplement in cell and microbial culture media

TRUSTED_SUSTAINABILITY

Specifications

Name Albumin
pH Range 6 to 8
Quantity 50 g
Source Egg
Packaging Glass Bottle
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.