Learn More
Thermo Scientific Chemicals Albumin, for biochemistry, from chicken egg, powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
• It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa
Applications
• BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
• It is used to stabilize enzymes in restriction digestions in molecular biology experiments
• It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
• BSA is a nutritive agent and used as a supplement in cell and microbial culture media
Specifications
Specifications
| Name | Albumin |
| pH Range | 6 to 8 |
| Quantity | 50 g |
| Source | Egg |
| Packaging | Glass Bottle |
| Form | Powder |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.