Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture Grade, Low Endotoxin

Catalog No. AAJ65097A1
Click to view available options
Quantity:
10 g
50 g
100 g
1 kg

Albumin bovine/fraction V, CAS # 9048-46-8, also known as bovine serum albumin (BSA), is a serum albumin protein derived from cows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Albumin bovine/fraction V is a serum albumin protein commonly used in laboratory experiments including, enzyme-linked immunosorbent assay (ELISAs), immunoblots and immune-histochemistry or -fluorescence

Applications

• Albumin bovine/fraction V displays blocking activity mainly when casein-sensitive antibodies, including phospho-specific antibodies, are used
• In genetic approaches this compound can be used to increase polymerase chain reactions (PCR) yields from low purity templates
• This compound is useful for stabilizing enzymes during storage and also for enzymatic reactions where the absence of nucleases is crucial
• In in vitro cultures, albumin bovine/fraction V has been applied as a nutrient in cell and microbial cultures especially when serum-free media is used
• It is commonly used as a protein standard to obtain calibration curves for several laboratorial experiments
• Albumin bovine/fraction V is a small protein commonly used in immunohistochemistry to block nonspecific regions. The binding of this compound to nonspecific binding sites increases the probability of the antibodies binds only to the region of interest.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized powder
Quantity 1 kg
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture grade, Low Endotoxin
Recombinant Native
Form Lyophilized
Purity or Quality Grade Cell Culture grade, 98%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.