Learn More
MP Biomedicals™ Albumin, Chicken, MP Biomedicals™
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g
Description
From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa. Differential scanning calorimetry indicates that chicken egg albumin denatures at 84°C.
- Can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen
- Useful as a marker (∼45 kDa) for protein electrophoresis
- Has been used to produce an animal model of asthma
- Used as a medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil
- Was used in a study testing intestinal macromolecule absorption in the fetal pig after infusion of colostrum in utero
Specifications
Specifications
| Quantity | 5 g |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov