Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALCAM/CD166 Antibody (001), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290895AF647
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Rat samples. It is validated for Western Blot, ELISA.Specifications
ALCAM/CD166 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human ALCAM No cross-reactivity in ELISA with: Human BCAM | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
1 | |
Western Blot, ELISA | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ALCAM/CD166 (O35112; Met1-Lys527). | |
0.1 mL | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
214 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction