Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALCAM/CD166 Antibody (200622) [Biotin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1172B
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
ALCAM/CD166 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
200622 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166, Trp28-Lys527, Accession # AAC06342 | |
0.1 mL | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
214 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction