Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALCAM/CD166 Antibody (21B9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ALCAM/CD166 |
---|---|
Clone | 21B9 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
ALCAM/CD166 | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
A synthesized peptide derived from Human ALCAM/CD166 [UniProt Q13740] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
21B9 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
214 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title