Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Alcare Plus™ Antiseptic Handrub with Emollients

Catalog No. p-2916602
Click to view available options
Volume (Metric):
160 mL
266 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Fast-acting, foamed, alcohol-based handwash with added emollients that moisturize skin

Ideal for use when soap and water are not available.

  • Contains 62% ethyl alcohol by volume
  • Provides broad spectrum activity against pathogenic microorganisms, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE)
  • Spreads easily to facilitate even distribution over all skin surfaces for effective disinfection

Specifications

Certifications/Compliance FDA
For Use With (Application) Provides broad spectrum activity against pathogenic microorganisms, kills 99.99% of mrsa and vre in 5 sec.
Volume (Metric) 266 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.