Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Alcian Blue 8GX

Catalog No. p-7041754
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6012209 10 g
AAJ6012214 25 g
AAJ6012222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6012209 Supplier Thermo Scientific Chemicals Supplier No. J6012209
Only null left
Add to Cart

CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol

Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.

Solubility
Soluble in water, ethanol, ethylene glycol and 2-ethoxyethanol. Insoluble in xylene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33864-99-2
Molecular Formula C56H68Cl4CuN16S4
Molecular Weight (g/mol) 1298.86
MDL Number MFCD00010720
InChI Key XRWQTDIELZDCPS-UHFFFAOYSA-J
Synonym Ingrain blue 1; C.I. 74240
PubChem CID 17749099
IUPAC Name (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C

Specifications

Melting Point 148°C
Merck Index 14,218
Solubility Information Soluble in water,ethanol,ethylene glycol and 2-ethoxyethanol; Insoluble in xylene.
Formula Weight 1298.88
Color Blue
Quantity 10 g
Physical Form Powder
Chemical Name or Material Alcian Blue 8GX
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 251-705-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.