Learn More
Epredia™ Alcian Blue/PAS Special Stain Kit
Catalog No. p-4530172 Shop All Epredia Products
Click to view available options
Type:
Alcian Blue/PAS Special Stain Kit
This item is not returnable. View return policy
Catalog No. 23-900-664
Supplier: Epredia™ 87023
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Demonstrate the presence of both neutral and acidic mucosubstances with the Epredia™ Alcian Blue/PAS Special Stain Kit
  • Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
  • Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances
Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent
Specifications
Type Alcian Blue/PAS Special Stain Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.