Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alcohol, 190 Proof, BiotechGrade, 95-96%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Alcohol, 190 Proof, BiotechGrade is an extremely volatile, flammable, colorless liquid, pure ethyl alcohol (ethanol) that are ideal for applications in industrial, health, education, and research. Biotech grade is a reliable, high-quality, reagent for general laboratory use and is commonly used in thermometers, as fuel and as a powerful solvent. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absence of inhibitors.
Specifications
Specifications
CAS Max % | 100% |
UN Number | UN1170 |
Percent Purity | 95 to 96% |
Grade | Biotech |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction