Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, 190 Proof, USP, 94.9-96%, Spectrum™ Chemical
SDP

Catalog No. 18600368
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum

ET108, 64-17-5, C2H5OH

Spectrum™ Chemical Alcohol, 190 Proof, USP is approximately 95% Ethanol and 5% water. This product, 190 proof USP grade alcohol, is used as an antiseptic to disinfect skin prior to injections and as a solvent. All Spectrum Chemical USP products, including all USP alcohol products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
Specific Gravity 0.812-0.816
UN Number UN1170
Percent Purity 94.9 to 96%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.