Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5383232 Shop All Avantor Products
Click to view available options
Quantity:
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle
Poly Bottle

High quality chemicals for laboratory and specialized industrial use

    For Histological Use; NOTE: Intended for nonfuel end purposes only

Chemical Identifiers

CAS 64-17-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name methanol
SMILES CO

Specifications

Quantity 4 L
Flash Point 13.00°C (55.4°F)
Packaging Aluminum Bottle
Specific Gravity 1L = 0.79kg
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Alcohol, Anhydrous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.