Learn More
Alcohol, Anhydrous, Denatured, 200 Proof, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Spectrum™ Chemical Alcohol, Anhydrous, Denatured, 200 Proof is ethanol that has added additives that render it undrinkable, bad tasting or even poisonous to discourage people from consuming it. The additives do not chemically change the alcohol and thus it is identical to a normal alcohol apart from it's smell, color and taste and can be used for the exact same purposes as normal alcohol. Typical denaturants include methyl alcohol, isopropyl alcohol and methyl isobutyl ketone.
Specifications
Specifications
| CAS Max % | 85.8%, 4.3%, 8.96%, 0.91% |
| Quantity | 200 L |
| Packaging | Fiber Drum |
| UN Number | UN1987 |
| Percent Purity | 84 to 88% |
| Grade | Ungraded |
| Chemical Name or Material | Alcohol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.