Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alcohol, Anhydrous, Reagent, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
1 L
19 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth Polyethylene Bottle
Poly Pail
Description
- For Histological Use
Specifications
Specifications
| Quantity | 500 mL |
| Flash Point | 13.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 0.79kg |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Alcohol, Anhydrous, Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction