Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alcohol Dehydrogenase 1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $371.50
Specifications
Antigen | Alcohol Dehydrogenase 1A |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145381
|
Invitrogen™
PA5145381 |
200 μL |
Each for $371.50
|
|
|||||
FEPA5145382
|
Invitrogen™
PA5145382 |
20 μL |
Each for $191.80
|
|
|||||
Description
This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.Specifications
Alcohol Dehydrogenase 1A | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Pig | |
P06757, P07327 | |
124, 24172 | |
Recombinant Human ADH1 protein, Lys169-Arg370 (Accession #P07327). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ADC1; Adh; ADH, alpha subunit; ADH1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; alcohol dehydrogenase A subunit; alcohol dehydrogenase ADH1; Alcohol dehydrogenase I; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; O0947; YADH-1; YOL086C | |
Adh1, ADH1A | |
IgG | |
Protein A | |
ADH1A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title