Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alcohol dehydrogenase 1B Antibody (OTI4F12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270092F
Description
alcohol dehydrogenase 1B Monoclonal specifically detects alcohol dehydrogenase 1B in Human, Rat, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
alcohol dehydrogenase 1B | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ADH1B | |
Full length recombinant protein of human alcohol dehydrogenase 1B (NP_000659) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Canine | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4F12 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ADH, beta subunit, ADH2alcohol dehydrogenase 1B, alcohol dehydrogenase 1B (class I), beta polypeptide, alcohol dehydrogenase 2 (class I), beta polypeptide, Alcohol dehydrogenase subunit beta, aldehyde reductase, DKFZp686C06125, EC 1.1.1, EC 1.1.1.1 | |
Mouse | |
Affinity Purified | |
Cancer | |
125.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction