Learn More
Invitrogen™ Alcohol Dehydrogenase 1B Monoclonal Antibody (OTI5D7)

Description
Alcohol Dehydrogenase 1B Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | Alcohol Dehydrogenase 1B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5D7 |
| Concentration | 0.69 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ADH1B |
| Gene Accession No. | P00325 |
| Gene Alias | 2700007F14Rik; acetaldehyde dehydrogenase 5; ADH, beta subunit; ADH1B; ADH2; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase subunit beta; aldehyde dehydrogenase 1 family member B1; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; aldehyde dehydrogenase X, mitochondrial; aldehyde reductase; ALDH class 2; Aldh1b1; ALDH5; Aldhx; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.