Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alcohol Dehydrogenase 1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597435
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597435 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597435 Supplier Invitrogen™ Supplier No. PA597435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Alcohol Dehydrogenase 1B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by this gene is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This encoded protein, consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alcohol Dehydrogenase 1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ADH1B
Gene Accession No. P00325
Gene Alias 2700007F14Rik; acetaldehyde dehydrogenase 5; ADH, beta subunit; ADH1B; ADH2; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase subunit beta; aldehyde dehydrogenase 1 family member B1; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; aldehyde dehydrogenase X, mitochondrial; aldehyde reductase; ALDH class 2; Aldh1b1; ALDH5; Aldhx; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117
Gene Symbols ADH1B
Host Species Rabbit
Immunogen Recombinant Human Alcohol dehydrogenase 1B protein (2-375AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.