Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alcohol dehydrogenase 4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP19023325UL
Description
alcohol dehydrogenase 4 Polyclonal specifically detects alcohol dehydrogenase 4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
alcohol dehydrogenase 4 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
P08319 | |
ADH4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ALGATDCLNPRDLHKPIQEVIIELTKGGVDFALDCAGGSETMKAALDCTTAGWGSCTFIGVAAGSKGLTVFPEELIIGRTINGTFFGGWKSVDSIPKLVTDYKNKKFNLDALV | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ADH-2, alcohol dehydrogenase 4, alcohol dehydrogenase 4 (class II), pi polypeptide, Alcohol dehydrogenase class II pi chain, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 | |
Rabbit | |
40 kDa | |
25 μL | |
Lipid and Metabolism | |
127 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction