Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alcohol dehydrogenase 5 Antibody (OTI5G4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270096H
Description
alcohol dehydrogenase 5 Monoclonal specifically detects alcohol dehydrogenase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
alcohol dehydrogenase 5 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ADH5 | |
Recombinant protein fragment corresponding to amino acids 1-266 of human alcohol dehydrogenase 5 (NP_000662) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5G4 | |
Western Blot, Immunohistochemistry | |
ADH-3, ADHXEC 1.1.1.1, alcohol dehydrogenase (class III), chi polypeptide, Alcohol dehydrogenase 5, alcohol dehydrogenase 5 (class III), chi polypeptide, Alcohol dehydrogenase class chi chain, alcohol dehydrogenase class-3, Alcohol dehydrogenase class-III, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.284, FALDH, FDHGSNOR, formaldehyde dehydrogenase, Glutathione-dependent formaldehyde dehydrogenase, GSH-FDH, S-(hydroxymethyl)glutathione dehydrogenase | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
128.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction