Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alcohol dehydrogenase 7 Antibody (OTI2H10), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270098V
Description
alcohol dehydrogenase 7 Monoclonal specifically detects alcohol dehydrogenase 7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
alcohol dehydrogenase 7 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADH7 | |
Full length recombinant protein of human alcohol dehydrogenase 7 (NP_000664) produced in HEK293T cells. | |
0.1 mL | |
131.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2H10 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
ADH4, ADH-4, alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide, alcohol dehydrogenase class 4 mu/sigma chain, Alcohol dehydrogenase class IV mu/sigma chain, alcohol dehydrogenase VII, alcohol dehydrogenase-7, class IV sigma-1 alcohol dehydrogenase, class IV sigmasigma alcohol dehydrogenase, EC 1.1.1, EC 1.1.1.1, Gastric alcohol dehydrogenase, Retinol dehydrogenase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction