Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alcohol dehydrogenase Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518332
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518332 Supplier Invitrogen™ Supplier No. PA518332
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alcohol dehydrogenase Pan
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADH1B
Gene Accession No. P00325, P00326, P07327
Gene Alias ADH beta; ADH gamma; ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 1C; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117; pan ADH
Gene Symbols ADH1A, ADH1B, ADH1C
Host Species Goat
Immunogen Synthetic peptide sequence (STAGKVMKCKA) corresponding to the N-terminus amino acids of ADH1A, B, C.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124, 125, 126
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.