Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol dehydrogenase Pan Recombinant Rabbit Monoclonal Antibody (1X3C0)
GREENER_CHOICE

Catalog No. PIMA542716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542716 Supplier Thermo Scientific Supplier No. MA542716
Add to Cart
Add to Cart
This item is not returnable. View return policy

Alcohol dehydrogenase Pan Recombinant Rabbit Monoclonal Antibody (1X3C0)

Alcohol dehydrogenase Pan Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Alcohol dehydrogenase Pan
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1X3C0
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADH1B
Gene Accession No. P00325, P00326, P07327
Gene Alias ADH beta; ADH gamma; ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 1C; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117; pan ADH
Gene Symbols ADH1A, ADH1B, ADH1C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Alcohol Dehydrogenase (P07327).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124, 125, 126
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.