Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alcohol Dehydrogenase (Yeast)

Catalog No. 89933082 Shop All Abnova Corporation Products
Click to view available options
Quantity:
100 mg

Used for Enzyme

#REF!

Specifications

Accession Number P25377
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 149.5kDa
Name Alcohol Dehydrogenase (Yeast)
Preparation Method Native protein purified from Yeast
Quantity 100 mg
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name ALCOHOL
Assays Enzymatic determination of primary alcohols, and aldehydes; Synthesis of chiral compounds; Spectrophotometric assay of plasmalogenase; Enzymatic catalysis in organic solvents; Studies of NAD+, NADH, NADP+, and NADPH.
Biological Activity ≥300 units per mg protein. One Unit reduces one micromole of NAD per minute at 25°C, pH 8.8.
Species Yeast
Protein Tag None
Expression System Native protein purified from Yeast
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.