Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol DehydrogenaseRabbit anti-Yeast, Polyclonal, Rockland™
SDP

Catalog No. 501991019
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1019 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1019 Supplier Rockland Immunochemicals Supplier No. 20041430100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-ALCOHOL DEHYDROGENASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Alcohol Dehydrogenase [Yeast]. Cross reactivity against Alcohol Dehydrogenase from other sources may occur but have not been specifically determined.

This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.

Specifications

Antigen Alcohol Dehydrogenase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Adh1
Gene Accession No. P00330
Gene Alias Adh; ADH, alpha subunit; Adh1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; Alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; Alcohol dehydrogenase A subunit; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase
Gene Symbols Adh1
Host Species Rabbit
Immunogen Alcohol Dehydrogenase [Yeast]
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854068
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.