Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access BNP Calibrators

Catalog No. 21280116 Shop All Beckman Coulter Products
Click to view available options
:
Access Bnp Calibrators
This item is not returnable. View return policy
This item is not returnable. View return policy

Access BNP assay offers exceptional clinical quality for cardiac testing.

  • B-type natriuretic peptide (BNP) levels are related to the severity of signs and symptoms of heart failure
  • Access BNP is the only BNP assay on automated immunoassay platforms with two risk stratification claims FDA cleared for patients with Heart Failure and patients with Acute Coronary Syndrome
  • Outstanding Analytical Sensitivity: 1 pg/mL
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes BNP
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Disposable Limited-use
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.