Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Reagent ACS, 94 to 96%, MilliporeSigma™

Catalog No. p-3176616
Click to view available options
Quantity:
500 mL
4 L
208 L
Packaging:
Glass Bottle
Poly Bottle
Poly-coated Glass Bottle
Steel Barrel

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
SMILES CCO

Specifications

Color Colorless
Quantity 500 mL
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range Assay (ethanol and methanol): 94.0 to 96.0% (v/v); Assay (ethanol): 89.0 to 92.0% (v/v); Assay (isopropyl alcohol): 4.0 to 6.0% (v/v); Assay (methanol): 3.5 to 5.5% (v/v)
Residue after Evaporation ≤0.001%
Linear Formula CH3CH2OH with methanol and isopropyl alcohol
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide
IUPAC Name Ethanol
Percent Purity 94 to 96%
Grade ACS Reagent
Chemical Name or Material Alcohol
Show More Show Less
Recommended Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.