Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcoholic Sulfuric Acid, For Phosphorus, LabChem™

Catalog No. LC105702 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC105702 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC105702 Supplier LabChem, Inc. Supplier No. LC105702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-93-9 , 67-56-1
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
CAS Max % 98, 2
Linear Formula H2SO4
UN Number UN2924
Solubility Information Soluble in water
Formula Weight 98.08
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Carbon monoxide Carbon dioxide May release flammable gases
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Alcoholic Sulfuric Acid, For Phosphorus
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer (inhalation).
Causes damage to organs (central nervous system, kidney, liver, optic nerve).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Use only outdoors or in a well-ventilated area.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor/physician.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep at rest in a position comfortable for breathing.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry extinguishing powder, alcohol-resistant foam, carbon dioxide to extinguish.
Wash contaminated clothing before reuse.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.