Learn More
Alconox™ Tergajet™ Low-Foaming Phosphate-Free Powdered Detergent

Description
Tergajet is a highly alkaline detergent containing sodium carbonate, silicate of soda, and surfactants.
- Concentrated, biodegradable and disposable
- Corrosion inhibited formula
- Free rinsing, no interfering residues
- Passes inhibitory residue test
- Dilute 1:100 for use (10g in 1L of water)
- Ideal for under-counter and floor standing lab washing machines
Used to clean:
Water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning compliant.
Used to remove:
Soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.
Surfaces cleaned:
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for Aluminum.
Cleaning method:
Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-in place and manual cleaning if proper skin and eye protection is used.
Shelf life: 2 years from date of manufacture.
Order Info
3.8L (1 gal.) makes up to 98.4L (26 gal.) cleaning solution
Specifications
Specifications
Product Type | Low-Foaming Phosphate-Free Powdered Detergent |
Form | Powder |
pH | 11 |
Appearance | White |
Container Type | Box |
Content And Storage | Store closed and upright in a cool dry place at 15° to 30°C. |
For Use With (Application) | Enzyme cleaner for use in labware washers, manual and ultrasonic cleaning |
Volume (English) | 4 lb. |
Volume (Metric) | 1.8 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.