Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Tergajet™ Low-Foaming Phosphate-Free Powdered Detergent
GREENER_CHOICE

Catalog No. p-4939754 Shop All Alconox Products
Click to view available options
Volume (English):
4 lb.
25 lb.
50 lb.
300 lb.
Container Type:
Box
Carton
Drum
This item is not returnable. View return policy

Catalog No. 16-000-159

Supplier: Alconox™ 2203

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nonionic, heavy duty enzyme cleaner for use in labware washers, manual and ultrasonic cleaning

Tergajet is a highly alkaline detergent containing sodium carbonate, silicate of soda, and surfactants.

  • Concentrated, biodegradable and disposable
  • Corrosion inhibited formula
  • Free rinsing, no interfering residues
  • Passes inhibitory residue test
  • Dilute 1:100 for use (10g in 1L of water)
  • Ideal for under-counter and floor standing lab washing machines

Used to clean:
Water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning compliant.

Used to remove:
Soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.

Surfaces cleaned:
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for Aluminum.

Cleaning method:
Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-in place and manual cleaning if proper skin and eye protection is used.

Shelf life: 2 years from date of manufacture.

Order Info

3.8L (1 gal.) makes up to 98.4L (26 gal.) cleaning solution

Specifications

Product Type Low-Foaming Phosphate-Free Powdered Detergent
Form Powder
pH 11
Appearance White
Container Type Drum
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Enzyme cleaner for use in labware washers, manual and ultrasonic cleaning
Volume (English) 300 lb.
Volume (Metric) 136 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.