Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ AlcoSCRUB™ Instant Antiseptic Hand Cleanser Squeeze Bottle
Shop All Epredia Products
Click to view available options
Volume (English):
4 oz.
16 oz.
32 oz.
Packaging:
Pump bottle
Squeeze Bottle
Description
- Meets CDC guidelines
- Formulated with emollients to be gentle on skin
- 99.99% effective against a broad spectrum of microorganisms
Specifications
Specifications
| Product Type | Antiseptic Hand Wash |
| Description | Pump bottle |
| Packaging | Pump bottle |
| Volume (English) | 32 oz. |
| Volume (Metric) | 946 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction