Learn More
Description
- Allows concurrent use of other red fluorescent reporters or probes enables flow cytometry-based detection of intracellular Aldehyde Dehydrogenase (ALDH) enzyme activity
- High ALDH activity serves as a universal marker of stem cells, both normal and malignant
- AldeGreen™ Reagent: 1 vial containing 50 μg of AldeGreen™ reagent powder
- DEAB Reagent: 1 vial containing 1 mL of DEAB (Diethylaminobenzaldehyde) reagent
- Hydrochloric Acid (2N): 1 vial containing 1.5 mL of 2 Normal hydrochloric acid
- Dimethylsulphoxide: 1 vial containing 1.5 mL of Dimethylsulphoxide (DMSO)
- AldeGreen™ Assay Buffer: 4 bottles containing 25 mL of AldeGreen™ Assay Buffer each
- Verapamil: 4 vials containing 615 μg Verapamil powder each
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C, Reconstituted and activated may be stored in aliquots (at -20 °C protected from light for up to 1 year, Not recommended repeated freezing and thawing) |
| Detection Method | Fluorescence |
| For Use With (Application) | Flow Cytometry, Fluorescence Activated Cell Sorting (FACS) |
| Quantity | 1 Kit |
| Excitation Wavelength | 515 to 545 nm |
| Emission Wavelength | 488 |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.