Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (023), DyLight 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289893UV
Description
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
Monoclonal | |
DyLight 350 | |
acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 (NP_000680.2; Ser 2-Ser 501). | |
0.1 mL | |
Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
216 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
23 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction