Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH16A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988158
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8158 Supplier Bioss Supplier No. BS7956R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen ALDH16A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ALDH16A1
Gene Accession No. Q8IZ83
Gene Alias 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; Aldehyde dehydrogenase family 16 member A1; ALDH16A1; BOS_17636; mFLJ00209
Gene Symbols ALDH16A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ALDH16A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126133
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.