Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (JE62-68)
GREENER_CHOICE

Catalog No. PIMA544816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544816 Supplier Invitrogen™ Supplier No. MA544816
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ALDH16A1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH16A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE62-68
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ALDH16A1
Gene Accession No. Q3T1L0, Q571I9, Q8IZ83
Gene Alias 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209
Gene Symbols ALDH16A1
Host Species Rabbit
Immunogen Synthetic peptide within human ALDH16A1 aa 550-600/802.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126133, 361571, 69748
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.