Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH18A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552546 Supplier Invitrogen™ Supplier No. PA552546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIVEQVSVLQ NQGREMMLVT SGAVAFGKQR LRHEILLSQS VRQALHSGQN QLKEMAIPVL EARACAAAGQ SGLMALYEAM FTQYSICAAQ ILVTNLDFHD EQKRRNLNGT LHELLRMNIV PIVNTNDAVV PPAEPNSDLQ GVNVISVKD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH18A1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALDH18A1
Gene Accession No. P54886, Q9Z110
Gene Alias 2810433K04Rik; ADCL3; AI429789; aldehyde dehydrogenase 18 family member A1; aldehyde dehydrogenase 18 family, member A1; aldehyde dehydrogenase 18A1; aldehyde dehydrogenase family 18 member A1; aldh18a1; ARCL3A; cb842; cb899; delta1-pyrroline-5-carboxlate synthetase; delta-1-pyrroline-5-carboxylate synthase; delta-1-pyrroline-5-carboxylate synthetase; Gamma-glutamyl kinase; Gamma-glutamyl phosphate reductase; GK; Glutamate 5-kinase; glutamate gamma-semialdehyde synthetase; Glutamate-5-semialdehyde dehydrogenase; Glutamyl-gamma-semialdehyde dehydrogenase; GPR; GSAS; MGC117316; P5CS; Pycs; pyrroline-5-carboxylate synthetase; pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase); sb:cb881; similar to pyrroline-5-carboxylate synthetase isoform 1; SPG9A; SPG9B; wu:fa91f10; wu:fi05f11; wu:fi17d12
Gene Symbols ALDH18A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALDH18A1. Recombinant protein control fragment (Product #RP-89746).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361755, 56454, 5832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.